missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-1-(2-furylmethyl)piperidine hydrochloride, 95%, Thermo Scientific™

Product Code. 10597664
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10597664 250 mg Amber Glass Bottle 250mg
10507674 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10597664 Supplier Thermo Scientific Maybridge Supplier No. CC68574CB
 Restricted Item
Estimated Shipment: 12-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 944450-92-4
Molecular Formula C11H17Cl2NO
Molecular Weight (g/mol) 250.163
MDL Number MFCD10700059
InChI Key SRRFZUHMIKVVPV-UHFFFAOYSA-N
Synonym 4-chloromethyl-1-2-furylmethyl piperidine hydrochloride, 4-chloromethyl-1-furan-2-ylmethyl piperidine hydrochloride, 2-4-chloromethyl piperidyl methyl furan, chloride, 4-chloromethyl-1-furan-2-yl methyl piperidine-hydrogen chloride 1/1
PubChem CID 43811048
IUPAC Name 4-(chloromethyl)-1-(furan-2-ylmethyl)piperidine;hydrochloride
SMILES C1CN(CCC1CCl)CC2=CC=CO2.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 250.17
Percent Purity 95%
Chemical Name or Material 4-(Chloromethyl)-1-(2-furylmethyl)piperidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.