accessibility menu, dialog, popup

UserName

CAS RN 94497-51-5

IUPAC Name: 4-[(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl]benzoic acid
Synonyms: tamibarotene amnoid tamibaro retinobenzoic acid
Molecular Weight (g/mol): 351.45
Molecular Formula: C22H25NO3
InChi Key: MUTNCGKQJGXKEM-UHFFFAOYSA-N
SMILES: CC1(C)CCC(C)(C)C2=CC(NC(=O)C3=CC=C(C=C3)C(O)=O)=CC=C12
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)