accessibility menu, dialog, popup

UserName

CAS RN 95-13-6

CAS RN 95-13-6

IUPAC Name: 1H-indene
Synonyms: indene indonaphthene
Molecular Weight (g/mol): 116.16
Molecular Formula: C9H8
InChi Key: YBYIRNPNPLQARY-UHFFFAOYSA-N
SMILES: C1C=CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (7)
  • (7)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (6)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (3)
  • (7)
  • (7)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)