accessibility menu, dialog, popup

UserName

CH3NH2H3C

CAS RN 95-64-7

IUPAC Name: 3,4-dimethylaniline
Synonyms: 3,4-dimethylaniline 3,4-dimethylaminobenzene
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: DOLQYFPDPKPQSS-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C=C1C
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)