missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylaniline, TRC

Product Code. 30376217
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30376217 5 g 5g
30370820 25 g -
30369187 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30376217 Supplier Toronto Research Chemicals Supplier No. TRCD4621855G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-64-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
Synonym 3,4-Xylidine (8CI), 3,4-Dimethylbenzenamine, 1-Amino-3,4-dimethylbenzene, 3,4-Dimethylaminobenzene, 3,4-Dimethylaniline, 3,4-Dimethylphenylamine, 3,4-Xylylamine, 4-Amino-1,2-dimethylbenzene, 4-Amino-o-xylene, NSC 41800, NSC 7099, 3,4-Dimethylaniline, 3,4-Xylidine, 1-Amino-3,4-dimethylbenzene, 3,4-Dimethylaminobenzene, 3,4-Dimethylaniline, 3,4-Dimethylbenzenamine, 3,4-Dimethylphenylamine, 3,4-Xylylamine, 4-Amino-1,2-dimethylbenzene, 4-Amino-o-xylene, NSC 41800, NSC 7099
IUPAC Name 3,4-dimethylaniline
SMILES Cc1ccc(N)cc1C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H11N/c1-6-3-4-8(9)5-7(6)2/h3-5H,9H2,1-2H3
Formula Weight 121.0891
Chemical Name or Material 3,4-Dimethylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.