accessibility menu, dialog, popup

UserName

H3COOCH3OOOH

CAS RN 95110-10-4

IUPAC Name: 2-(2,6-dimethoxyphenoxy)acetic acid
Synonyms: 2,6-dimethoxyphenoxyacetic acid
Molecular Weight (g/mol): 212.2
Molecular Formula: C10H12O5
InChi Key: MCUFTNXSWDHDIS-UHFFFAOYSA-N
SMILES: COC1=CC=CC(OC)=C1OCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results