CAS RN 954-81-4
IUPAC Name:
2-(5-bromopentyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms:
2-(5-bromopentyl)isoindole-1,3-dione
Molecular Weight (g/mol):
296.16
Molecular Formula:
C13H14BrNO2
InChi Key:
QKVHAKICMNABGB-UHFFFAOYSA-N
SMILES:
BrCCCCCN1C(=O)C2=CC=CC=C2C1=O
1
–
1
of
1
results
1
N-(5-Bromopentyl)phthalimide, 97%
CAS: 954-81-4 Molecular Formula: C13H14BrNO2 Molecular Weight (g/mol): 296.164 MDL Number: MFCD00060522 InChI Key: QKVHAKICMNABGB-UHFFFAOYSA-N Synonym: n-5-bromopentyl phthalimide,2-5-bromopentyl isoindole-1,3-dione,n-5-bromopentyl-phthalimide,2-5-bromopentyl isoindoline-1,3-dione,2-5-bromo-pentyl-isoindole-1,3-dione,2-5-bromopentyl-1h-isoindole-1,3 2h-dione,2-5-bromopentyl-2,3-dihydro-1h-isoindole-1,3-dione,5-phthalimidopentyl bromide,1-bromo-5-phthalimidopentane PubChem CID: 136770 IUPAC Name: 2-(5-bromopentyl)isoindole-1,3-dione SMILES: C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCBr