missing translation for 'onlineSavingsMsg'
Learn More

N-(5-Bromopentyl)phthalimide, 97%

Product Code. 11388486
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11388486

Brand: Thermo Scientific Alfa Aesar L17543.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 954-81-4 | C13H14BrNO2 | 296.164 g/mol

It is a pharmaceutical intermediate and is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate and is used in medicine.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 954-81-4
Molecular Formula C13H14BrNO2
Molecular Weight (g/mol) 296.164
MDL Number MFCD00060522
InChI Key QKVHAKICMNABGB-UHFFFAOYSA-N
Synonym n-5-bromopentyl phthalimide, 2-5-bromopentyl isoindole-1,3-dione, n-5-bromopentyl-phthalimide, 2-5-bromopentyl isoindoline-1,3-dione, 2-5-bromo-pentyl-isoindole-1,3-dione, 2-5-bromopentyl-1h-isoindole-1,3 2h-dione, 2-5-bromopentyl-2,3-dihydro-1h-isoindole-1,3-dione, 5-phthalimidopentyl bromide, 1-bromo-5-phthalimidopentane
PubChem CID 136770
IUPAC Name 2-(5-bromopentyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCBr

Specifications

Melting Point 58°C to 62°C
Odor Odorless
Quantity 100 g
Beilstein 177015
Solubility Information Insoluble in water.
Formula Weight 296.17
Percent Purity 97%
Chemical Name or Material N-(5-Bromopentyl)phthalimide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.