accessibility menu, dialog, popup

UserName

NH2NNNN(R)(S)OH(R)OHO

CAS RN 958-09-8

IUPAC Name: (2R,3S,5R)-5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Synonyms: 2-deoxyadenosine
Molecular Weight (g/mol): 251.25
Molecular Formula: C10H13N5O3
InChi Key: OLXZPDWKRNYJJZ-RRKCRQDMSA-N
SMILES: NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)