missing translation for 'onlineSavingsMsg'
Learn More

2’-Deoxyadenosine, TRC

Product Code. 30367298
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367298 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367298 Supplier Toronto Research Chemicals Supplier No. TRCD2395835G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2'-Deoxyadenosine
CAS 958-09-8
Purity Grade Notes HPLC
Molecular Formula C10 H13 N5 O3
Quantity 5 g
Synonym 167: PN: US20040053876 SEQID: 167 unclaimed DNA, 9-(2-Deoxy-β-D-erythro-pentofuranosyl)adenine, NSC 141848, NSC 143510, NSC 83258, 1-(6-Amino-9H-purin-9-yl)-1,2-dideoxy-β-D-ribofuranose
InChI Formula InChI=1S/C10H13N5O3/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(17)6(2-16)18-7/h3-7,16-17H,1-2H2,(H2,11,12,13)/t5-,6+,7+/m0/s1
SMILES Nc1ncnc2c1ncn2[C@H]3C[C@H](O)[C@@H](CO)O3
IUPAC Name (2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular Weight (g/mol) 251.242
Formula Weight 251.102
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.