accessibility menu, dialog, popup

UserName

CAS RN 959236-97-6

H3CSNNBr

CAS RN 959236-97-6

IUPAC Name: 4-bromo-2-(methylsulfanyl)pyrimidine
Synonyms: 4-bromo-2-(methylsulfanyl)pyrimidine
Molecular Weight (g/mol): 205.07
Molecular Formula: C5H5BrN2S
InChi Key: TZSQAGYDWPPKPE-UHFFFAOYSA-N
SMILES: CSC1=NC=CC(Br)=N1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results