accessibility menu, dialog, popup

UserName

CAS RN 959996-58-8

NSOOClFFF

CAS RN 959996-58-8

IUPAC Name: 6-(trifluoromethyl)pyridine-3-sulfonyl chloride
Synonyms: 6-(trifluoromethyl)pyridine-3-sulfonyl chloride
Molecular Weight (g/mol): 245.6
Molecular Formula: C6H3ClF3NO2S
InChi Key: NSGLNIQAWVNZFB-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC=C(C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)