missing translation for 'onlineSavingsMsg'
Learn More

6-(Trifluoromethyl)pyridine-3-sulfonyl chloride, 97%

Product Code. 15488745
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488745 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15488745 Supplier Thermo Scientific Alfa Aesar Supplier No. H34033.03
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 959996-58-8 | C6H3ClF3NO2S | 245.6 g/mol

6-Trifluoromethyl-3-pyridinesulfonyl Chloride is a chemical used for the preparation of N-sulfonamido polycyclic pyrazolyl compounds for treatment of cognitive disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Trifluoromethyl-3-pyridinesulfonyl Chloride is a chemical used for the preparation of N-sulfonamido polycyclic pyrazolyl compounds for treatment of cognitive disorders.

Notes
Store in cool place. Keep container ti
TRUSTED_SUSTAINABILITY

Specifications

CAS 959996-58-8
Melting Point 42°C to 45°C
Molecular Formula C6H3ClF3NO2S
MDL Number MFCD10699112
Quantity 1 g
UN Number UN3261
Synonym 6-trifluoromethyl pyridine-3-sulfonyl chloride, 6-trifluoromethyl-3-pyridinesulfonyl chloride, 3-pyridinesulfonylchloride,6-trifluoromethyl, chloro 6-trifluoromethyl 3-pyridyl sulfone, 3-pyridinesulfonyl chloride, 6-trifluoromethyl
InChI Key NSGLNIQAWVNZFB-UHFFFAOYSA-N
SMILES C1=CC(=NC=C1S(=O)(=O)Cl)C(F)(F)F
IUPAC Name 6-(trifluoromethyl)pyridine-3-sulfonyl chloride
Molecular Weight (g/mol) 245.6
PubChem CID 45594311
Formula Weight 245.61
Percent Purity 97%
Chemical Name or Material 6-(Trifluoromethyl)pyridine-3-sulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.