accessibility menu, dialog, popup

UserName

CAS RN 96-29-7

CH3CH3NHO(E/Z)

CAS RN 96-29-7

IUPAC Name: (E)-N-(butan-2-ylidene)hydroxylamine
Synonyms: 2-butanone oxime methylethyl ketone oxime
Molecular Weight (g/mol): 87.12
Molecular Formula: C4H9NO
InChi Key: WHIVNJATOVLWBW-SNAWJCMRSA-N
SMILES: CC\C(C)=N\O
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)