missing translation for 'onlineSavingsMsg'
Learn More

2-Butanone oxime, 99%

Product Code. 11318035
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318035 1000 mL 1 L
11398025 100 mL 100mL
11308035 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11318035 Supplier Thermo Scientific Alfa Aesar Supplier No. L08196.AU
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 96-29-7 | C4H9NO | 87.12 g/mol

2-Butanone oxime was employed as reagent and solvent in the syntheses of ketoimine and 2,4-dipyridyl-1,3,5-triazapentadiene palladium(II) complexes. It was also used in the synthesis of novel acetaldiimine cobalt complex, [CoI2{((CH3CH2)(CH3)C=NO)2C(CH3)2}]. Methyl ethyl ketoxime is used primarily as an antiskinning agent in alkyd coating resins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butanone oxime was employed as reagent and solvent in the syntheses of ketoimine and 2,4-dipyridyl-1,3,5-triazapentadiene palladium(II) complexes. It was also used in the synthesis of novel acetaldiimine cobalt complex, [CoI2{((CH3CH2)(CH3)C=NO)2C(CH3)2}]. Methyl ethyl ketoxime is used primarily as an antiskinning agent in alkyd coating resins

Solubility
Soluble in water at 20°C 114 g/L. Soluble in most organic solvents .

Notes
Store away from acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-29-7
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00013935
InChI Key WHIVNJATOVLWBW-SNAWJCMRSA-N
Synonym mek-oxime, butanone oxime, methyl ethyl ketone oxime, 2-butanone oxime, ethyl methyl ketoxime, usaf ek-906, ethyl-methylketonoxim, unii-51yge935u9, 2-butanone, oxime, methyl ethyl ketoxime
PubChem CID 5324276
IUPAC Name (NZ)-N-butan-2-ylidenehydroxylamine
SMILES CC\C(C)=N\O

Specifications

Melting Point -30°C
Density 0.921
Boiling Point 152°C to 153°C
Flash Point 62°C (144°F)
Refractive Index 1.443
Quantity 1000 mL
Beilstein 1698241
Solubility Information Soluble in water at 20°C 114g/L. Soluble in most organic solvents .
Formula Weight 87.12
Percent Purity 99%
Chemical Name or Material 2-Butanone oxime
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.