CAS RN 96839-26-8
CAS RN 96839-26-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,4-Diaminobutane-1,4-13C2 Dihydrochloride, TRC
CAS: 96839-26-8 Molecular Formula: C213C2H12N2 . 2(HCl) Molecular Weight (g/mol): 163.06 Synonym: 1,4-Butanediamine-1,4-13C2, dihydrochloride (9CI),1,4-Butanediamine-1,4-13C2 dihydrochloride,1,4-Butanediamine-13C2 dihydrochloride,1,4-Diaminobutane-1,4-13C2 dihydrochloride,Putrescine-13C2 dihydrochloride SMILES: Cl.Cl.N[13CH2]CC[13CH2]N