missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diaminobutane-1,4-13C2 Dihydrochloride, TRC

Product Code. 30352241
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30352241 10 mg 10mg
30349850 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30352241 Supplier Toronto Research Chemicals Supplier No. TRCD41602910MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96839-26-8
Molecular Formula C213C2H12N2 . 2(HCl)
Molecular Weight (g/mol) 163.06
Synonym 1,4-Butanediamine-1,4-13C2, dihydrochloride (9CI), 1,4-Butanediamine-1,4-13C2 dihydrochloride, 1,4-Butanediamine-13C2 dihydrochloride, 1,4-Diaminobutane-1,4-13C2 dihydrochloride, Putrescine-13C2 dihydrochloride
SMILES Cl.Cl.N[13CH2]CC[13CH2]N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C4H12N2.2ClH/c5-3-1-2-4-6;;/h1-6H2;2*1H/i3+1,4+1;;
Formula Weight 162.0601
Chemical Name or Material 1,4-Diaminobutane-1,4-13C2 Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.