accessibility menu, dialog, popup

UserName

ClSOOBr

CAS RN 98-58-8

IUPAC Name: 4-bromobenzene-1-sulfonyl chloride
Synonyms: 4-bromobenzenesulfonyl chloride
Molecular Weight (g/mol): 255.51
Molecular Formula: C6H4BrClO2S
InChi Key: KMMHZIBWCXYAAH-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)