missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzenesulfonyl chloride, 98+%

Product Code. 11468436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468436 500 g 500g
11448436 25 g 25g
11458436 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468436 Supplier Thermo Scientific Alfa Aesar Supplier No. A15909.36
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 98-58-8 | C6H4BrClO2S | 255.51 g/mol

4-Bromobenzenesulfonyl chloride is used as activating agent in the synthesis of oligodeoxyribo- and oligoribo- nucleotides in solution. It also used in the synthesis of 4-(N-allylsulfamoyl)phenylboronic acid and in protection of amines as 4-bromobenzenesulfonamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzenesulfonyl chloride is used as activating agent in the synthesis of oligodeoxyribo- and oligoribo- nucleotides in solution. It also used in the synthesis of 4-(N-allylsulfamoyl)phenylboronic acid and in protection of amines as 4-bromobenzenesulfonamides.

Solubility
Soluble in Tetrahydrofuran, petroleum ether and 1,4-dioxane. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-58-8
Molecular Formula C6H4BrClO2S
Molecular Weight (g/mol) 255.51
MDL Number MFCD00007437
InChI Key KMMHZIBWCXYAAH-UHFFFAOYSA-N
Synonym 4-bromobenzene-1-sulfonyl chloride, p-bromobenzenesulfonyl chloride, 4-bromo-benzenesulfonyl chloride, p-bromophenylsulfonyl chloride, benzenesulfonyl chloride, 4-bromo, 4-bromobenzenesulphonyl chloride, 4-bromobenzenesulfonylchloride, unii-c6347t8fp1, 4-bromobenzene sulfonyl chloride, 4-bromophenyl chlorosulfone
PubChem CID 7396
IUPAC Name 4-bromobenzenesulfonyl chloride
SMILES C1=CC(=CC=C1S(=O)(=O)Cl)Br

Specifications

Melting Point 73°C to 77°C
Boiling Point 152°C to 154°C (26 mmHg)
Quantity 500 g
UN Number UN3261
Beilstein 743518
Sensitivity Moisture sensitive
Merck Index 14,1407
Solubility Information Soluble in Tetrahydrofuran,petroleum ether and 1,4-dioxane. Insoluble in water.
Formula Weight 255.52
Percent Purity ≥98%
Chemical Name or Material 4-Bromobenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.