accessibility menu, dialog, popup

UserName

CH3H3CCH3OONO

CAS RN 98977-36-7

IUPAC Name: tert-butyl 3-oxopiperidine-1-carboxylate
Synonyms: tert-butyl 3-oxopiperidine-1-carboxylate
Molecular Weight (g/mol): 199.25
Molecular Formula: C10H17NO3
InChi Key: RIFXIGDBUBXKEI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCCC(=O)C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results