missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-3-piperidone, 97%

Product Code. 10294534 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10294534 25 g Glass bottle 25g
10153344 5 g Glass bottle 5g
10113064 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10294534 Supplier Thermo Scientific Acros Supplier No. 397000250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 98977-36-7 | C10H17NO3 | 199.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98977-36-7
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD01631193
InChI Key RIFXIGDBUBXKEI-UHFFFAOYSA-N
Synonym 1-boc-3-piperidone, 1-boc-3-piperidinone, n-boc-3-piperidone, 1-n-boc-3-piperidone, n-boc-3-piperidinone, boc-3-piperidone, 1-tert-butoxycarbonyl-3-piperidone, 3-oxo-piperidine-1-carboxylic acid tert-butyl ester, 3-oxopiperidine, n-boc protected, tert-butyl 3-oxopiperidinecarboxylate
PubChem CID 2756825
IUPAC Name tert-butyl 3-oxopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(=O)C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 37°C to 39°C
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 199.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-BOC-3-piperidone
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-3-piperidone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.