accessibility menu, dialog, popup

UserName

CAS RN 99-28-5

OHBrBrNOO

CAS RN 99-28-5

IUPAC Name: 2,6-dibromo-4-nitrophenol
Synonyms: 2,6-dibromo-4-nitrophenol
Molecular Weight (g/mol): 296.9
Molecular Formula: C6H3Br2NO3
InChi Key: WBHYZUAQCSHXCT-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(C=C1Br)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results