accessibility menu, dialog, popup

UserName

CAS RN 99-97-8

IUPAC Name: N,N,4-trimethylaniline
Synonyms: dimethyltolylamine
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: GYVGXEWAOAAJEU-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C)C=C1
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results