accessibility menu, dialog, popup

UserName

ClHSnH3ClHClHH3C

CAS RN 993-16-8

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 246.14
Molecular Formula: CH9Cl3Sn
InChi Key: VLOZTKJSAQVQRJ-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.C[SnH3]
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results