accessibility menu, dialog, popup

UserName

CAS RN 99783-23-0

CH3FFFOHO

CAS RN 99783-23-0

IUPAC Name: 4,4,4-trifluoro-2-methylbutanoic acid
Synonyms: 4,4,4-trifluoro-2-methylbutanoic acid
Molecular Weight (g/mol): 156.1
Molecular Formula: C5H7F3O2
InChi Key: OBUKEMPWKGDRJV-UHFFFAOYNA-N
SMILES: CC(CC(F)(F)F)C(O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results