Glycine, ≥99%, Molecular Biology Grade, Ultrapure
CAS: 56-40-6 | C2H5NO2 | 75.067 g/mol
Additional Details:
CAS Number:
56-40-6
Weight:
1.00000kg
| Quantity | 1 kg |
|---|---|
| Packaging | Plastic Bottle |
Disclaimers:
RUO – Research Use Only
Chemical Identifiers
| CAS | 56-40-6 |
|---|---|
| Molecular Formula | C2H5NO2 |
| Molecular Weight (g/mol) | 75.067 |
| MDL Number | MFCD00008131 |
| InChI Key | DHMQDGOQFOQNFH-UHFFFAOYSA-N |
| Synonym | glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine |
| PubChem CID | 750 |
| ChEBI | CHEBI:15428 |
| IUPAC Name | 2-aminoacetic acid |
| SMILES | C(C(=O)O)N |
Specification
| Chemical Name or Material | Glycine |
|---|---|
| Melting Point | ∼245°C (decomposition) |
| CAS | 56-40-6 |
| Molecular Formula | C2H5NO2 |
| Packaging | Plastic Bottle |
| Linear Formula | H2NCH2CO2H |
| MDL Number | MFCD00008131 |
| Beilstein | 635782 |
| Synonym | glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine |
| Synonym | glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine |
| Synonym | glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine |
| Synonym | glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine |
| Synonym | glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine |
| Synonym | glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine |
| Synonym | glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine |
| Synonym | glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine |
| Synonym | glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine |
| Synonym | glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine |
| InChI Key | DHMQDGOQFOQNFH-UHFFFAOYSA-N |
| Density | 1.595 |
| SMILES | C(C(=O)O)N |
| IUPAC Name | 2-aminoacetic acid |
| Molecular Weight (g/mol) | 75.067 |
| Quantity | 1 kg |
| PubChem CID | 750 |
| ChEBI | CHEBI:15428 |
| Formula Weight | 75.07 |
| Percent Purity | ≥99% |
| Specific Gravity | 1.595 |
| Grade | Molecular Biology |
| Physical Form | Powder |