missing translation for 'onlineSavingsMsg'
Learn More

Glycine, ≥99%, Molecular Biology Grade, Ultrapure

Product Code. p-7082820 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15815158 500 g Plastic Bottle 500g
15835298 1 kg Plastic Bottle 1kg
15845298 5 kg Plastic Bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15815158 Supplier Thermo Scientific Supplier No. J16407.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-40-6 | C2H5NO2 | 75.067 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.067
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES C(C(=O)O)N

Specifications

Melting Point ∼245°C (decomposition)
Packaging Plastic Bottle
Linear Formula H2NCH2CO2H
Beilstein 635782
Density 1.595
Quantity 500 g
Formula Weight 75.07
Percent Purity ≥99%
Specific Gravity 1.595
Grade Molecular Biology
Physical Form Powder
Chemical Name or Material Glycine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.