missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-2-(2,4,5-Trichlorophenoxy)propionic-3,3,3-d3 Acid, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Fenoprop-D3 (propanoic-3,3,3-D3) |
| Molecular Formula | C9 D3 H4 Cl3 O3 |
| Quantity | 0.01 g |
| Synonym | (±)-2-(2,4,5-Trichlorophenoxy)propionic-3,3,3-d3 Acid, Fenoprop-d3, Fenoprop-D3 (propanoic-3,3,3-D3), 2-(2,4,5-Trichlorophenoxy)propanoic-3,3,3-d3 acid |
| InChI Formula | InChI=1 S/C9H7Cl3O3/c1-4(9(13)14)15-8-3-6(11)5(10)2-7(8)12/h2-4 H,1H3,(H,13,14)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(Oc1cc(Cl)c(Cl)cc1Cl)C(=O)O |
| IUPAC Name | 3,3,3-trideuterio-2-(2,4,5-trichlorophenoxy)propanoic acid |
| Molecular Weight (g/mol) | 272.5276 |
| Formula Weight | 270.9649 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction