missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4-Trichlorobiphenyl-2',3',4',5',6'-d5, CDN
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 2,3,4-Trichlorobiphenyl-2',3',4',5',6'-D5 |
| CAS | 1219805-70-5 |
| Molecular Formula | C12 D5 H2 Cl3 |
| Quantity | 0.01 g |
| Synonym | 2',3',4'-Trichloro-1,1'-biphenyl-2,3,4,5,6-d5, 2,3,4-Trichloro-1,1'-biphenyl-2',3',4',5',6'-d5, 2,3,4-Trichlorobiphenyl-2',3',4',5',6'-D5, PCB 21-d5, 2,3,4-TriCB-2',3',4',5',6'-d5 |
| InChI Formula | InChI=1 S/C12H7Cl3/c13-10-7-6-9(11(14)12(10)15)8-4-2-1-3-5-8/h1-7 H/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2ccc(Cl)c(Cl)c2Cl |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-(2,3,4-trichlorophenyl)benzene |
| Molecular Weight (g/mol) | 262.5738 |
| Formula Weight | 260.9927 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction