missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4'-Dichlorobiphenyl-d8, CDN
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 2,4'-Dichlorobiphenyl-D8 |
| CAS | 1219795-19-3 |
| Molecular Formula | C12 D8 Cl2 |
| Quantity | 0.01 g |
| Synonym | 2,4'-Dichloro-1,1'-biphenyl-2',3,3',4,5,5',6,6'-d8, 2,4'-Dichlorobiphenyl-D8, CB 8-d8, PCB 8-d8, 2,4'-DiCB-d8 |
| InChI Formula | InChI=1 S/C12H8Cl2/c13-10-7-5-9(6-8-10)11-3-1-2-4-12(11)14/h1-8 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c(Cl)c1[2 H])c2c([2 H])c([2 H])c(Cl)c([2 H])c2[2 H] |
| IUPAC Name | 1-chloro-2-(4-chloro-2,3,5,6-tetradeuteriophenyl)-3,4,5,6-tetradeuteriobenzene |
| Molecular Weight (g/mol) | 231.1472 |
| Formula Weight | 230.0505 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction