missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Methyl-d3-thio)propylamine, CDN
Click to view available options
Quantity:
0.01 g
0.025 g
Unit Size:
0.01g
0.03g
Specifications
Specifications
| Chemical Name or Material | 3-(Methylthio)propylamine-D3 (S-methyl-D3) |
| CAS | 2256713-59-2 |
| Molecular Formula | C4 D3 H8 N S |
| Quantity | 0.01 g |
| Synonym | 1-Propanamine, 3-(methyl-d3-thio)- (ACI), 3-(Methyl-d3-thio)-1-propanamine (ACI), 3-(Methylthio)propylamine-d3, 3-(Methylthio)propylamine-D3 (S-methyl-D3), 3-(Trideuteriomethylthio)-1-propanamine, 3-Methyl-d3-sulfanyl-1-propylamine |
| InChI Formula | InChI=1 S/C4H11NS/c1-6-4-2-3-5/h2-5H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])SCCCN |
| IUPAC Name | 3-(trideuteriomethylsulfanyl)propan-1-amine |
| Molecular Weight (g/mol) | 108.2203 |
| Formula Weight | 108.0801 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction