missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylbutyryl-DL-carnitine-d9 HCl (N,N,N-trimethyl-d9), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Isovaleryl-DL-carnitine-D9 (N,N,N-trimethyl-D9) Chloride |
| CAS | 2608872-54-2 |
| Molecular Formula | C12 D9 H15 N O4 . Cl |
| Quantity | 0.01 g |
| Synonym | Isovaleryl-DL-carnitine-d9 HCl (N,N,N-tri-methyl-d9), Isovaleryl-DL-carnitine-d9 Chloride, Isovaleryl-DL-carnitine-D9 (N,N,N-trimethyl-D9) Chloride, 3-Carboxy-N,N,N-tri(methyl-d3)-2-(3-methylbutanoyloxy)-1-propanaminium chloride, [3-Carboxy-2-(3-methylbutanoyloxy)propyl]tri(methyl-d3)ammonium chloride |
| InChI Formula | InChI=1 S/C12H23NO4.ClH/c1-9(2)6-12(16)17-10(7-11(14)15)8-13(3,4)5;/h9-10 H,6-8H2,1-5H3;1 H/i3D3,4D3,5D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](CC(CC(=O)O)OC(=O)CC(C)C)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | [3-carboxy-2-(3-methylbutanoyloxy)propyl]-tris(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 290.8317 |
| Formula Weight | 290.1959 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction