missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pregnen-11beta,21-diol-3,20-dione-2,2,4,6,6,17alpha,21,21-d8, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Corticosterone-d8 |
| CAS | 1271728-07-4 |
| Molecular Formula | C21 2H8 H22 O4 |
| Quantity | 0.01 g |
| Synonym | Pregn-4-ene-3,20-dione-2,2,4,6,6,17,21,21-d8, 11,21-dihydroxy-, (11β)-, Corticosterone-d8, Corticosterone-2,2,4,6,6,17,21,21-D8, (11β)-11,21-Dihydroxypregn-4-ene-3,20-dione-2,2,4,6,6,17,21,21-d8 |
| InChI Formula | InChI=1 S/C21H30O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h9,14-17,19,22,24 H,3-8,10-11H2,1-2H3/t14-,15-,16+,17-,19+,20-,21-/m0/s1/i3D2,7D2,9 D,11D2,16 D |
| SMILES | [2 H]C1=C2C([2 H])([2 H])C[C@H]3 [C@@H]4 CC[C@]([2 H])(C(=O)C([2 H])([2 H])O)[C@@]4(C)C[C@H](O)[C@@H]3 [C@@]2(C)CC([2 H])([2 H])C1=O |
| IUPAC Name | (8 S,9 S,10 R,11 S,13 S,14 S,17 S)-2,2,4,6,6,17-hexadeuterio-17-(2,2-dideuterio-2-hydroxyacetyl)-11-hydroxy-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-1 H-cyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 354.51 |
| Formula Weight | 354.2646 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction