missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pregnen-6alpha-methyl-17alpha-ol-3,20-dione-2,2,4,6,21,21,21-d7 Acetate, CDN
Click to view available options
Quantity:
0.005 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Medroxyprogesterone Acetate-D7 (2,2,4,6-D4,acetyl-D3) |
| Molecular Formula | C24 D7 H27 O4 |
| Quantity | 0.005 g |
| Synonym | 4-Pregnen-6 alpha-methyl-17 alpha-ol-3,20-dione-2,2,4,6,21,21,21-d7 Acetate, (6α)-17-(Acetyl-2,2,2-d3-oxy)-6-methylpregn-4-ene-3,20-dione-2,2,4,6-d4, Medroxy progesterone 17-acetate-d7, Medroxyprogesterone Acetate-D7 (2,2,4,6-D4,acetyl-D3) |
| InChI Formula | InChI=1 S/C24H34O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h13-14,18-20 H,6-12H2,1-5H3/t14-,18+,19-,20-,22+,23-,24-/m0/s1/i3D3,6D2,13 D,14 D |
| SMILES | [2 H]C1=C2[C@@]([2 H])(C)C[C@@H]3 [C@H](CC[C@@]4(C)[C@H]3 CC[C@]4(OC(=O)C([2 H])([2 H])[2 H])C(=O)C)[C@@]2(C)CC([2 H])([2 H])C1=O |
| IUPAC Name | [(6 S,8 R,9 S,10 R,13 S,14 S,17 R)-17-acetyl-2,2,4,6-tetradeuterio-6,10,13-trimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-1 H-cyclopenta[a]phenanthren-17-yl] 2,2,2-trideuterioacetate |
| Molecular Weight (g/mol) | 393.5675 |
| Formula Weight | 393.2896 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction