missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5alpha-Androstan-3alpha,17beta-diol-16,16,17-d3, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | d3-5 A-Androstane-3 A,17 b-diol |
| CAS | 79037-33-5 |
| Molecular Formula | C19 2H3 H29 O2 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C19H32O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12-17,20-21 H,3-11H2,1-2H3/t12-,13+,14-,15-,16-,17-,18-,19-/m0/s1/i6D2,17 D |
| SMILES | [2 H]C1([2 H])C[C@H]2 [C@@H]3 CC[C@H]4 C[C@H](O)CC[C@]4(C)[C@H]3 CC[C@]2(C)[C@@]1([2 H])O |
| IUPAC Name | (3 R,5 S,8 R,9 S,10 S,13 S,14 S,17 S)-16,16,17-trideuterio-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15-dodecahydro-1 H-cyclopenta[a]phenanthrene-3,17-diol |
| Molecular Weight (g/mol) | 295.475 |
| Formula Weight | 295.259 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction