missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5alpha-Cholestan-3alpha-ol-2,2,3,4,4-d5, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Epicholestanol-2,2,3,4,4-D5 |
| Molecular Formula | C27 D5 H43 O |
| Quantity | 0.005 g |
| Synonym | (3α,5α)-Cholestan-3-ol-2,2,3,4,4-d5, 5α-Cholestan-3α-ol-2,2,3,4,4-d5, α-Cholestanol-2,2,3,4,4-d5, epi-Cholestanol-d5, Epicholestanol-2,2,3,4,4-D5, (3α,5α)-Cholestanol-2,2,3,4,4-d5 |
| InChI Formula | InChI=1 S/C27H48O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h18-25,28 H,6-17H2,1-5H3/t19-,20+,21-,22+,23-,24+,25+,26+,27-/m1/s1/i13D2,17D2,21 D |
| SMILES | [2 H]C1([2 H])C[C@]2(C)[C@H]3 CC[C@]4(C)[C@H](CC[C@H]4 [C@@H]3 CC[C@H]2 C([2 H])([2 H])[C@]1([2 H])O)[C@H](C)CCCC(C)C |
| IUPAC Name | (3 R,5 S,8 R,9 S,10 S,13 R,14 S,17 R)-2,2,3,4,4-pentadeuterio-10,13-dimethyl-17-[(2 R)-6-methylheptan-2-yl]-1,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-ol |
| Molecular Weight (g/mol) | 393.7002 |
| Formula Weight | 393.4019 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction