missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5alpha-Pregnan-21-ol-3,20-dione-2,2,4,4,17alpha,21,21-d7, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 5 Alpha-Dihydrodeoxycorticosterone-d7 |
| Molecular Formula | C21H25D7O3 |
| Quantity | 0.005 g |
| Synonym | 21-Hydroxy-5α-pregnane-3,20-dione-d7, 5α-Dihydro-11-deoxycorticosterone-d7, 5α-Pregnan-21-ol-3,20-dione-d7, Allopregnane-21-ol-3,20-dione-d7 |
| InChI Formula | InChI=1 S/C21H32O3/c1-20-9-7-14(23)11-13(20)3-4-15-16-5-6-18(19(24)12-22)21(16,2)10-8-17(15)20/h13,15-18,22 H,3-12H2,1-2H3/t13-,15-,16-,17-,18+,20-,21-/m0/s1/i7D2,11D2,12D2,18 D |
| SMILES | [H][C@@]12 CC[C@@]3([H])C([2 H])([2 H])C(C([2 H])([2 H])C[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4([2 H])C(C([2 H])([2 H])O)=O)=O |
| IUPAC Name | (5 S,8 R,9 S,10 S,13 S,14 S,17 S)-17-(2-hydroxyacetyl-d2)-10,13-dimethylhexadecahydro-3 H-cyclopenta[a]phenanthren-3-one-2,2,4,4,17-d5 |
| Molecular Weight (g/mol) | 339.52 |
| Formula Weight | 339.28 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction