missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7beta-Hydroxycholesterol-25,26,26,26,27,27,27-d7, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 7 beta-Hydroxycholesterol 25,26,26,26,27,27,27-d7 |
| CAS | 349553-97-5 |
| Molecular Formula | C27 2H7 H39 O2 |
| Quantity | 0.005 g |
| Synonym | Cholest-5-ene-25,26,26,26,27,27,27-d7-3,7-diol, (3β,7β)-, (3β,7β)-Cholest-5-ene-25,26,26,26,27,27,27-d7-3,7-diol, 7β-Hydroxycholesterol-25,26,26,26,27,27,27-D7, (3β,7β)-Cholest-5-ene-3,7-diol-d7, 7β-Hydroxycholesterol-d7 |
| InChI Formula | InChI=1 S/C27H46O2/c1-17(2)7-6-8-18(3)21-9-10-22-25-23(12-14-27(21,22)5)26(4)13-11-20(28)15-19(26)16-24(25)29/h16-18,20-25,28-29 H,6-15H2,1-5H3/t18-,20+,21-,22+,23+,24+,25+,26+,27-/m1/s1/i1D3,2D3,17 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(CCC[C@@H](C)[C@H]1 CC[C@H]2 [C@@H]3 [C@@H](O)C=C4C[C@@H](O)CC[C@]4(C)[C@H]3 CC[C@]12 C)C([2 H])([2 H])[2 H] |
| IUPAC Name | (3 S,7 R,8 S,9 S,10 R,13 R,14 S,17 R)-10,13-dimethyl-17-[(2 R)-6,7,7,7-tetradeuterio-6-(trideuteriomethyl)heptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthrene-3,7-diol |
| Molecular Weight (g/mol) | 409.7 |
| Formula Weight | 409.3937 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction