missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agomelatine-d3 (acetamide-2,2,2-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Agomelatine-d3 (acetamide-2,2,2-d3) |
| CAS | 1079389-38-0 |
| Molecular Formula | C15 D3 H14 N O2 |
| Quantity | 0.005 g |
| Synonym | Acetamide-2,2,2-d3, N-[2-(7-methoxy-1-naphthalenyl)ethyl]- (ACI), N-[2-(7-Methoxy-1-naphthalenyl)ethyl]acetamide-2,2,2-d3 (ACI), 2,2,2-Trideuterio-N-[2-(7-methoxynaphthalen-1-yl)ethyl]acetamide, Agomelatine-d3, Agomelatine-D3 (acetyl-D3), Agomelatine-d3 (acetamide-2,2,2-d3) |
| InChI Formula | InChI=1 S/C15H17NO2/c1-11(17)16-9-8-13-5-3-4-12-6-7-14(18-2)10-15(12)13/h3-7,10 H,8-9H2,1-2H3,(H,16,17)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)NCCc1cccc2ccc(OC)cc12 |
| IUPAC Name | 2,2,2-trideuterio-N-[2-(7-methoxynaphthalen-1-yl)ethyl]acetamide |
| Molecular Weight (g/mol) | 246.32 |
| Formula Weight | 246.145 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction