missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyanazine-d5 (N-ethyl-d5), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | CyanazineD5(N-ethylD5) |
| CAS | 1190003-29-2 |
| Molecular Formula | C9 2H5 H8 Cl N6 |
| Quantity | 0.005 g |
| Synonym | 2-[[4-Chloro-6-(1,1,2,2,2-pentadeuterioethylamino)-1,3,5-triazin-2-yl]amino]-2-methylpropanenitrile, Cyanazine D5 (N-ethyl D5), Cyanazine-d5 |
| InChI Formula | InChI=1 S/C9H13ClN6/c1-4-12-7-13-6(10)14-8(15-7)16-9(2,3)5-11/h4H2,1-3H3,(H2,12,13,14,15,16)/i1D3,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])Nc1nc(Cl)nc(NC(C)(C)C#N)n1 |
| IUPAC Name | 2-[[4-chloro-6-(1,1,2,2,2-pentadeuterioethylamino)-1,3,5-triazin-2-yl]amino]-2-methylpropanenitrile |
| Molecular Weight (g/mol) | 245.72 |
| Formula Weight | 245.1204 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction