missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmosterol-26,26,26,27,27,27-d6, CDN
missing translation for 'orderingAttributeHoverText'
Quantity:
0.005 g
0.01 g
missing translation for 'unitSize'
0.01g
Specifications
Specifications
| Chemical Name or Material | Desmosterol 26,26,26,27,27,27-d6 |
| CAS | 1246298-67-8 |
| Molecular Formula | C27 2H6 H38 O |
| Quantity | 0.005 g |
| Synonym | Cholesta-5,24-dien-26,26,26,27,27,27-d6-3-ol, (3β)-, (3β)-Cholesta-5,24-dien-26,26,26,27,27,27-d6-3-ol, Desmosterol-d6, Desmosterol-26,27-d6, Desmosterol-D6 (26,26,26,27,27,27-D6), 24,25-Dehydrocholesterol-26,27-d6, 24-Dehydrocholesterol-d6 |
| InChI Formula | InChI=1 S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h7,9,19,21-25,28 H,6,8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(=CCC[C@@H](C)[C@H]1 CC[C@H]2 [C@@H]3 CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3 CC[C@]12 C)C([2 H])([2 H])[2 H] |
| IUPAC Name | (3 S,8 S,9 S,10 R,13 R,14 S,17 R)-10,13-dimethyl-17-[(2 R)-7,7,7-trideuterio-6-(trideuteriomethyl)hept-5-en-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-3-ol |
| Molecular Weight (g/mol) | 390.67 |
| Formula Weight | 390.3769 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction