missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Desvenlafaxine-d6 Succinate H2O (N,N-dimethyl-d6), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (±)-Desvenlafaxine-d6 Succinate Hydrate (N,N-dimethyl-d6) |
| Molecular Formula | C16 D6 H19 N O2 . C4 H6 O4 . H2 O |
| Quantity | 0.005 g |
| Synonym | O-Desmethylvenlafaxine-D6 (N,N-dimethyl-D6) succinate monohydrate, 4-[2-[Di(methyl-d3)amino]-1-(1-hydroxycyclohexyl)ethyl]phenol butanedioic acid hydrate (1:1:1), D,L-O-Desmethyl venlafaxine-d6 succinate monohydrate, Desvenlafaxine-d6 succinate monohydrate, O-Desvenlafaxine-d6 succinate monohydrate, (±)-Desvenlafaxine-d6 Succinate H2O (N,N-dimethyl-d6) |
| InChI Formula | InChI=1 S/C16H25NO2.C4H6O4.H2O/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16;5-3(6)1-2-4(7)8;/h6-9,15,18-19 H,3-5,10-12H2,1-2H3;1-2H2,(H,5,6)(H,7,8);1H2/i1D3,2D3;; |
| SMILES | O.[2 H]C([2 H])([2 H])N(CC(c1ccc(O)cc1)C2(O)CCCCC2)C([2 H])([2 H])[2 H].OC(=O)CCC(=O)O |
| IUPAC Name | 4-[2-[bis(trideuteriomethyl)amino]-1-(1-hydroxycyclohexyl)ethyl]phenol;butanedioic acid;hydrate |
| Molecular Weight (g/mol) | 405.5155 |
| Formula Weight | 405.2634 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction