missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flecainide-d4 Acetate [bis(2,2,2-trifluoroethoxy-2,2-d2)], CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Flecainide-D4 (bis-trifluoroethoxy-D4) Acetate |
| CAS | 1276197-21-7 |
| Molecular Formula | C17 D4 H16 F6 N2 O3 . C2 H4 O2 |
| Quantity | 0.01 g |
| Synonym | Flecainide-d4 acetate, Flecainide-D4 (bis-trifluoroethoxy-D4) Acetate, N-(2-Piperidinylmethyl)-2,5-bis(2,2,2-trifluoroethoxy-1,1-d2)benzamide monoacetate, Flecainide-d4 Acetate [bis(2,2,2-trifluoroethoxy-1,1-d2)] |
| InChI Formula | InChI=1 S/C17H20F6N2O3.C2H4O2/c18-16(19,20)9-27-12-4-5-14(28-10-17(21,22)23)13(7-12)15(26)25-8-11-3-1-2-6-24-11;1-2(3)4/h4-5,7,11,24 H,1-3,6,8-10H2,(H,25,26);1H3,(H,3,4)/i8D2,9D2; |
| SMILES | [2 H]C([2 H])(NC(=O)c1cc(OC([2 H])([2 H])C(F)(F)F)ccc1OCC(F)(F)F)C2CCCCN2.CC(=O)O |
| IUPAC Name | acetic acid;N-[dideuterio(piperidin-2-yl)methyl]-5-(1,1-dideuterio-2,2,2-trifluoroethoxy)-2-(2,2,2-trifluoroethoxy)benzamide |
| Molecular Weight (g/mol) | 478.4193 |
| Formula Weight | 478.184 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction