missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lornoxicam-d4 (pyridine-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Lornoxicam-d4 |
| CAS | 1216527-48-8 |
| Molecular Formula | C13 2H4 H6 Cl N3 O4 S2 |
| Quantity | 0.005 g |
| InChI Formula | InChI=1 S/C13H10ClN3O4S2/c1-17-10(13(19)16-9-4-2-3-5-15-9)11(18)12-7(23(17,20)21)6-8(14)22-12/h2-6,18 H,1H3,(H,15,16,19)/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1nc(NC(=O)C2=C(O)c3sc(Cl)cc3S(=O)(=O)N2C)c([2 H])c([2 H])c1[2 H] |
| IUPAC Name | 6-chloro-4-hydroxy-2-methyl-1,1-dioxo-N-(3,4,5,6-tetradeuteriopyridin-2-yl)thieno[2,3-e]thiazine-3-carboxamide |
| Molecular Weight (g/mol) | 375.84 |
| Formula Weight | 375.0052 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction