missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Miconazole-d5 Nitrate (2,4-dichlorobenzyloxy-d5), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (±)-Miconazole-d5 Nitrate (2,4-dichlorobenzyloxy-d5) |
| CAS | 1398065-80-9 |
| Molecular Formula | C18 D5 H9 Cl4 N2 O . H N O3 |
| Quantity | 0.01 g |
| Synonym | (±)-Miconazole-d5 nitrate (2,4-dichlorobenzyloxy-d5), Miconazole-d5 nitrate, Miconazole-D5 (2,4-dichlorobenzyloxy-D5) nitrate, 1-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl-d3)methoxy-d2]ethyl]imidazole mononitrate, 1-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorobenzloxy-d5)ethyl]-1 H-imidazole nitrate |
| InChI Formula | InChI=1 S/C18H14Cl4N2O.HNO3/c19-13-2-1-12(16(21)7-13)10-25-18(9-24-6-5-23-11-24)15-4-3-14(20)8-17(15)22;2-1(3)4/h1-8,11,18 H,9-10H2;(H,2,3,4)/i1D,2 D,7 D,10D2; |
| SMILES | [2 H]c1c([2 H])c(c(Cl)c([2 H])c1Cl)C([2 H])([2 H])OC(Cn2ccnc2)c3ccc(Cl)cc3Cl.O[N+](=O)[O-] |
| IUPAC Name | 1-[2-(2,4-dichlorophenyl)-2-[dideuterio-(2,4-dichloro-3,5,6-trideuteriophenyl)methoxy]ethyl]imidazole;nitric acid |
| Molecular Weight (g/mol) | 484.172 |
| Formula Weight | 482.013 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction