missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(3-Methylcrotonyl)glycine-2,2-d2, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | N-(3-Methylcrotonyl)glycine-2,2-D2 |
| CAS | 1276197-31-9 |
| Molecular Formula | C7 D2 H9 N O3 |
| Quantity | 0.01 g |
| Synonym | N-(3-Methyl-1-oxo-2-buten-1-yl)glycine-2,2-d2, 2-(3-Methylbut-2-enamido)acetic-2,2-d2 acid, 3-Methylcrotonylglycine-2,2-d2, N-(3,3-Dimethylacrylyl)glycine-2,2-d2, β-Methylcrotonylglycine-d2, N-(3-Methylcrotonyl)glycine-2,2-d2 |
| InChI Formula | InChI=1 S/C7H11NO3/c1-5(2)3-6(9)8-4-7(10)11/h3H,4H2,1-2H3,(H,8,9)(H,10,11)/i4D2 |
| SMILES | [2 H]C([2 H])(NC(=O)C=C(C)C)C(=O)O |
| IUPAC Name | 2,2-dideuterio-2-(3-methylbut-2-enoylamino)acetic acid |
| Molecular Weight (g/mol) | 159.1795 |
| Formula Weight | 159.0864 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction