missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Aminobenzoyl-d4)glycine, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 4-Aminohippuric-d4 Acid |
| CAS | 1219805-41-0 |
| Molecular Formula | C9 2H4 H6 N2 O3 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C9H10N2O3/c10-7-3-1-6(2-4-7)9(14)11-5-8(12)13/h1-4 H,5,10H2,(H,11,14)(H,12,13)/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)NCC(=O)O)c([2 H])c([2 H])c1N |
| IUPAC Name | 2-[(4-amino-2,3,5,6-tetradeuteriobenzoyl)amino]acetic acid |
| Molecular Weight (g/mol) | 198.21 |
| Formula Weight | 198.0942 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction