missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl-d6-tetradecylamine N-Oxide, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethyltetradecylamine N-Oxide D6 (N,N-dimethyl-D6) |
| CAS | 2733717-74-1 |
| Molecular Formula | C16 D6 H29 N O |
| Quantity | 0.01 g |
| Synonym | N,N-Dimethyltetradecylamine N-oxide D6 (N,N-dimethyl-D6), N,N-Dimethyl-d6-tetradecylamine N-oxide, N,N-Di(methyl-d3)-1-tetradecylamine oxide, N,N-Di(methyl-d3)tetradecylamine N-oxide, Tetradecyldi(methyl-d3)amine N-oxide, Myristyldimethylamine-d6 oxide, N,N-Di(methyl-d3)-1-tetradecanamine N-oxide |
| InChI Formula | InChI=1 S/C16H35NO/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17(2,3)18/h4-16H2,1-3H3/i2D3,3D3 |
| SMILES | [2 H]C([2 H])([2 H])[N+]([O-])(CCCCCCCCCCCCCC)C([2 H])([2 H])[2 H] |
| IUPAC Name | N,N-bis(trideuteriomethyl)tetradecan-1-amine oxide |
| Molecular Weight (g/mol) | 263.492 |
| Formula Weight | 263.31 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction