missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic Acid-d15, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Specifications
Specifications
| Chemical Name or Material | N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic Acid D15 |
| CAS | 1219794-63-4 |
| Molecular Formula | C6 D15 N O6 S |
| Quantity | 0.01 g |
| Synonym | 2-[[2-(Hydroxy-d)-1,1-bis(hydroxy-d-methyl-d2)ethyl-2,2-d2]amino-d]ethanesulfonic-1,1,2,2,O-d5 acid, N-[2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]taurine-d15, 2-[[Tris(hydroxymethyl)methyl]amino]ethanesulfonic acid-15, N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic acid D15, TES-d15, Perdeutero-TES |
| InChI Formula | InChI=1 S/C6H15NO6S/c8-3-6(4-9,5-10)7-1-2-14(11,12)13/h7-10 H,1-5H2,(H,11,12,13)/i1D2,2D2,3D2,4D2,5D2,8 D,9 D,10 D/hD2 |
| SMILES | [2 H]OC([2 H])([2 H])C(N([2 H])C([2 H])([2 H])C([2 H])([2 H])S(=O)(=O)O[2 H])(C([2 H])([2 H])O[2 H])C([2 H])([2 H])O[2 H] |
| IUPAC Name | deuterio 1,1,2,2-tetradeuterio-2-[deuterio-[1,1,3,3-tetradeuterio-1,3-dideuteriooxy-2-[dideuterio(deuteriooxy)methyl]propan-2-yl]amino]ethanesulfonate |
| Molecular Weight (g/mol) | 244.344 |
| Formula Weight | 244.156 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction