missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nepsilon-Carboxymethyl-L-lysine-2,6,6-d3, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | N6-(Carboxymethyl)-L-lysine-2,6,6-D3 |
| CAS | 2699607-49-1 |
| Molecular Formula | C8 D3 H13 N2 O4 |
| Quantity | 0.01 g |
| Synonym | N6-(Carboxymethyl)-L-lysine-2,6,6-d3, N6-(Carboxymethyl)-L-Lysine-D3, NEpsilon-(1-Carboxymethyl)-L-lysine-d3, N6-(Carboxymethyl)-L-lysine-2,6,6-D3, (2 S)-2-Amino-6-(carboxymethylamino)hexanoic-2,6,6-d3 acid |
| InChI Formula | InChI=1 S/C8H16N2O4/c9-6(8(13)14)3-1-2-4-10-5-7(11)12/h6,10 H,1-5,9H2,(H,11,12)(H,13,14)/t6-/m0/s1/i4D2,6 D |
| SMILES | [2 H]C([2 H])(CCC[C@]([2 H])(N)C(=O)O)NCC(=O)O |
| IUPAC Name | (2 S)-2-amino-6-(carboxymethylamino)-2,6,6-trideuteriohexanoic acid |
| Molecular Weight (g/mol) | 207.242 |
| Formula Weight | 207.13 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction